missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-Rhamnose monohydrate, 98+%

Product Code. 11453227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453227 100 g 100g
11433227 5 g 5g
11443227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453227 Supplier Thermo Scientific Chemicals Supplier No. A16166.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol

L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

Solubility
Soluble in water and methanol. Slightly soluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10030-85-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00149363,MFCD00136036
InChI Key SHZGCJCMOBCMKK-HGVZOGFYSA-N
Synonym l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
PubChem CID 20849066
SMILES C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 89°C to 94°C
Density 1.47
Quantity 100 g
Beilstein 5988592
Merck Index 14,8172
Solubility Information Soluble in water and methanol. Slightly soluble in ethanol.
Optical Rotation +8° (c=10 in Water, 20h)
IUPAC Name (2R,3R,4R,5R,6S)-6-methyloxane-2,3,4,5-tetrol
Formula Weight 182.19 (164.17 Anhydrous)
Percent Purity 99%
Chemical Name or Material L-(+)-Rhamnose monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.