missing translation for 'onlineSavingsMsg'
Learn More

L-Rhamnose Monohydrate, TRC

Product Code. 30410621
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410621 100 g 100g
30425388 10 g 10g
30412031 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410621 Supplier Toronto Research Chemicals Supplier No. TRCR315000100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6155-35-7
Molecular Formula C6 H12 O5 . H2 O
Molecular Weight (g/mol) 182.17
Synonym α-L-Mannopyranose, 6-deoxy-, hydrate (1:1), Rhamnopyranose, monohydrate, α-L- (8CI), α-L-Mannopyranose, 6-deoxy-, monohydrate (9CI), α-L-Rhamnopyranose monohydrate, α-L-Rhamnose monohydrate, (2R,3R,4R,5R,6S)-2,3,4,5-Tetrahydroxy-6-methyltetrahydropyran monohydrate
IUPAC Name (2R,3R,4R,5R,6S)-6-methyloxane-2,3,4,5-tetrol;hydrate
SMILES O.C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H12O5.H2O/c1-2-3(7)4(8)5(9)6(10)11-2;/h2-10H,1H3;1H2/t2-,3-,4+,5+,6+;/m0./s1
Formula Weight 182.079
Chemical Name or Material L-Rhamnose, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.