Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Head Replacement Rotor Assembly
Product Code. 11898332 Shop All Masterflex Products
Change view
Click to view available options
For Use With:
Easy-Load II Pump Head 77200-50
Easy-Load II Pump Head 77200-52
Easy-Load II Pump Head 77200-62
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
11898332 Easy-Load II Pump Head 77200-52 Each
11725169 Easy-Load II Pump Head 77200-50 Each
11832603 Easy-Load II Pump Head 77200-62 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11898332 Supplier Masterflex™ Supplier No. 7720059
  • ONLINE EXCLUSIVE PRICE
    227.00 €
    207.87 € / Pack of 1
    Save 19.13 €
    8% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Masterflex™ L/S™ Easy-Load™ II Pump Head Replacement Rotor Assembly for use with High-Performance Precision Tubing
  • Get the most our of Masterflex™ L/S™ Easy-Load™ II Pump Heads with these replacement parts
  • Choose replacement parts suitable for your L/S™ pump head
  • Replacement of pump head rotor assembly extends the life of pump heads
Specifications
For Use With Easy-Load II Pump Head 77200-52
Material Cold-Rolled Steel
Product Line L/S
Tubing Type L/S™
Type Replacement Pump Head Rotor Assembly