missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Fixed-Speed Drive

Product Code. p-7184908
Click to view available options
Speed:
1 rpm
12 rpm
20 rpm
30 rpm
6 rpm
60 rpm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13025666

Brand: Masterflex™ 0754012

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Masterflex™ L/S™ Fixed-Speed Drive intended for reliable, general transfer and feed applications

  • Economical drives for reliable general transfer and feed applications
  • Painted Steel housed construction is IP 22 rated
  • Choose from seven speeds based on flow rate or number of heads required
  • Economy fixed-speed drives are stackable and ideal for lab, field, or process feed
  • Compact drives are ideal for applications where space is limited
  • Lighted power switch on the front of the drive indicates when the pump is running
  • Compatible with five different pump head types: Standard, Easy-Load™, Easy-Load™ II, Easy-Load™ 3, and Cartridge Pump Heads
  • Accepts up to 2 pump heads

Specifications

Amperage 0.5 amps
Certifications/Compliance Prop 65 Cancer and Repro Hazard
Control Type Fixed Speed
Dimensions (L x W x H) 152 x 134 x 118 mm
Electrical Requirements 115 VAC
Flow Rate 0.72 to 58 mL/min.
For Use With Standard, Easy-Load, Easy-Load II, Easy-Load 3, Cartridge Pump Heads
IP Rating IP22
Material Painted Steel
No. of Pump Heads 2 Pump Heads Accepted
Type Pump Drive
Speed 12 rpm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.