missing translation for 'onlineSavingsMsg'
Learn More

L-Serine benzyl ester hydrochloride, 98%

Product Code. 11412150
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11412150

Brand: Thermo Scientific Alfa Aesar H28790.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60022-62-0 | C10H14ClNO3 | 231.68 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60022-62-0
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.68
MDL Number MFCD00038955
InChI Key MGZWCDQAKCHOBX-UHFFFAOYNA-N
Synonym l-serine benzyl ester hydrochloride, h-ser-obzl.hcl, ser bzl .hcl, h-ser.obzl.hcl, benzyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, phenylmethyl ester, hydrochloride, l-serinebenzylesterhydrochloride, h-ser-obzl. hcl, pubchem19043, h-ser-obn hydrochloride
PubChem CID 12930233
SMILES Cl.NC(CO)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point ∽175°C (decomposition)
Solubility Information Soluble in water.
IUPAC Name benzyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
Quantity 1 g
Formula Weight 231.68
Optical Rotation −12° (c=4 in Water)
Percent Purity 98%
Chemical Name or Material L-Serine benzyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.