missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, MedChemExpress

Product Code. 16482539
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482539 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482539 Supplier Medchem Express Supplier No. HYN0650
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation.

L-Serine ((-)-Serine

Specifications

Chemical Name or Material L-Serine
CAS 56-45-1
Color White
Purity Grade Notes Research
Molecular Formula C3H7NO3
Synonym (-)-Serine (S)-Serine
Solubility Information H2O : 50 mg/mL (475.78 mM; Need ultrasonic)
SMILES N[C@@H](CO)C(O)=O
Molecular Weight (g/mol) 105.09
Quantity 1 g
Formula Weight 105.09
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.