missing translation for 'onlineSavingsMsg'
Learn More

L-Serine methyl ester hydrochloride, 98%

Product Code. 11499483
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499483 25 g 25g
11489483 5 g 5g
11409493 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499483 Supplier Thermo Scientific Alfa Aesar Supplier No. A12355.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5680-80-8 | C4H10ClNO3 | 155.578 g/mol

L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.

Solubility
Soluble in water (50 mg/ml), and methanol

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5680-80-8
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.578
MDL Number MFCD00063680
InChI Key NDBQJIBNNUJNHA-DFWYDOINSA-N
Synonym l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
PubChem CID 2723730
IUPAC Name methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point 155°C (decomposition)
Beilstein 3559591
Solubility Information Soluble in water (50mg/ml),and methanol
Quantity 25 g
Formula Weight 155.58
Optical Rotation +5° (c=2 in methanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Serine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.