missing translation for 'onlineSavingsMsg'
Learn More

L-Serine Methyl Ester Hydrochloride, TRC

Product Code. 30418745
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418745 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418745 Supplier Toronto Research Chemicals Supplier No. TRCS2710055G
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5680-80-8
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.58
Synonym (S)-Serine Methyl Ester Hydrochloride, Methyl L-serinate hydrochloride, L-Serine Methyl Ester Monohydrochloride
IUPAC Name methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CO

Specifications

InChI Formula InChI=1S/C4H9NO3.ClH/c1-8-4(7)3(5)2-6;/h3,6H,2,5H2,1H3;1H/t3-;/m0./s1
Quantity 5 g
Formula Weight 155.0349
Shipping Condition Room Temperature
Chemical Name or Material L-Serine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.