missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine methyl ester hydrochloride, 98%

Product Code. 10091430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091430 5 g Glass Bottle 5g
10513111 25 g Glass Bottle 25g
10540584 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091430 Supplier Thermo Scientific Chemicals Supplier No. 132670050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5680-80-8 | C4H10ClNO3 | 155.578 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5680-80-8
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.578
MDL Number MFCD00063680
InChI Key NDBQJIBNNUJNHA-DFWYDOINSA-N
Synonym l-serine methyl ester hydrochloride, h-ser-ome.hcl, methyl l-serinate hydrochloride, s-methyl 2-amino-3-hydroxypropanoate hydrochloride, methyl 2s-2-amino-3-hydroxypropanoate hydrochloride, l-serine, methyl ester, hydrochloride, l-methyl serinate hcl, serine methyl ester hydrochloride, l-serine methyl ester hcl, h-ser-ome-hcl
PubChem CID 2723730
IUPAC Name methyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl

Specifications

Melting Point 160.0°C to 165.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Beilstein 04, 506
Specific Rotation + 4.50
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Water 1% Max. (K.F.)
Specific Rotation Condition + 4.50 (20.00°C c=2,methanol)
Quantity 5 g
Formula Weight 155.58
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Serine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.