missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, TRC

Product Code. 17958304
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17958304 25 g 25g
17957924 1 g 1g
17958994 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17958304 Supplier Toronto Research Chemicals Supplier No. TRCS27099525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3 H7 N O3
Molecular Weight (g/mol) 105.09
Synonym Serine, (S)-2-amino-3-hydroxypropanoic acid,, (-)-Serine, (S)-2-Amino-3-hydroxypropanoic acid, (S)-Serine, (S)-α-Amino-β-hydroxypropionic acid, L-(-)-Serine, L-3-Hydroxy-2-aminopropionic acid, L-Ser, β-Hydroxy-L-alanine
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES N[C@@H](CO)C(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1
Quantity 25 g
Formula Weight 105.0426
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Serine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.