missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ L Shaped Spreader

Product Code. 17875588
Change view
Click to view available options
Quantity:
250
500
Unit Size:
Pack of 250
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17875588 500 Pack of 500
17845788 250 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17875588 Supplier Microspec™ Supplier No. E6G
  • ONLINE EXCLUSIVE PRICE
    59.39 €
    40.53 € / Pack of 500
    Save 18.86 €
    31% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Sterile L shaped inoculating spreaders

  • L Shaped Spreaders
  • 39 mm x 140 mm
  • Microseal Pouch
  • Sterile EtO

Specifications

Disposable Single-use
Autoclavable Not Autoclavable
Packaging Microseal Pouch of 5
Quantity 500
Size (English) 1.5 x 16.1 in.
Size (Metric) 39 x 410 mm
Material Plastic
Product Type Inoculating Spreader
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.