missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ L-Shaped Spreader

Product Code. 16445009
Change view
Click to view available options
Volume:
10 μL
Unit Size:
Pack of 6000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume unitSize
16445009 10 μL Pack of 6000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16445009 Supplier Wheaton™ Supplier No. 711315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Spreader calibrated to deliver a known volume of fluid sample to enable accurate microbiological analysis.

  • Meets quantitative standards for management of microbiological samples
  • L-shaped
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Volume 10 μL
Length (Metric) 172 mm
Material Polystyrene
Sterility Sterile
Color Green
Quantity 6000
Packaging Type Packaged in peel pouch
Calibrated Yes
Autoclavable Not Autoclavable
For Use With (Application) For accurate liquid transfer and plating
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.