missing translation for 'onlineSavingsMsg'
Learn More

L-Sorbose, TRC

Product Code. 17969264
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17969264 250 g 250g
17947724 50 g 50g
17937724 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17969264 Supplier Toronto Research Chemicals Supplier No. TRCS676875250G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-79-6
Molecular Formula C6 H12 O6
Molecular Weight (g/mol) 180.16
Synonym L-Sorbose, Sorbose, L- (8CI), Esorben, L-(-)-Sorbose, L-1,3,4,5,6-Pentahydroxyhexan-2-one, L-Sorbinose, L-xylo-2-Hexulose, NSC 97195, Sorbose
IUPAC Name (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES OC[C@H](O)[C@@H](O)[C@H](O)C(=O)CO

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5+,6+/m0/s1
Formula Weight 180.0634
Chemical Name or Material L(-)-Sorbose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.