missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid, 99+%

Product Code. 10081590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10081590 500 g Plastic bottle 500g
10655762 2.5 kg Plastic bottle 2.5kg
11905061 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081590 Supplier Thermo Scientific Acros Supplier No. 137855000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87-69-4 | C4H6O6 | 150.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 168.0°C to 172.0°C
Color White or Colorless
Quantity 500 g
Flash Point 210°C
Infrared Spectrum Authentic
Loss on Drying 0.5% (1g, 105°C) max.
Packaging Plastic bottle
Physical Form Crystals or Crystalline Powder
Assay Percent Range 99+%
Linear Formula HO2CCH(OH)CH(OH)CO2H
Solubility Information Solubility in water: 1390g/L (20°C). Other solubilities: soluble in methanol, ethanol, propanol and, glycerol, 4g/L ether, insoluble in chloroform
IUPAC Name 2,3-dihydroxybutanedioic acid
Formula Weight 150.09
Percent Purity ≥99%
Chemical Name or Material L(+)-Tartaric acid
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Tartaric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.