missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric acid, ACS

Product Code. 11387868
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387868 2 kg 2kg
11377868 100 g 100g
11397868 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387868 Supplier Thermo Scientific Alfa Aesar Supplier No. 036405.A3
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful as a resolving agent | CAS: 87-69-4 | C4H6O6 | 150.09 g/mol

L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and buffering agents. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and act as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in the farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

pH 2.1
Melting Point 168°C to 170°C
Beilstein 1725147
Merck Index 14,9070
Solubility Information Soluble in water,methanol and acetone.
IUPAC Name 2,3-dihydroxybutanedioic acid
Formula Weight 150.09
Grade ACS
Density 1.760 g/mL
Quantity 2 kg
Odor Odorless
Physical Form Granular
Chemical Name or Material L-(+)-Tartaric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.