missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Tartaric acid, disodium salt, dihydrate, 99+%, ACS reagent

Product Code. 10541804 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Unit Size:
100g
10kg
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10541804 100 g 100g
10269900 500 g 500g
10083863 2.5 kg 2.5kg
13468578 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10541804 Supplier Thermo Scientific Acros Supplier No. 424441000
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

pH 7.0 to 9.0 (5% soln. at 25°C)
Color White
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 15.61% to 15.71% (150°C)
Packaging Plastic bottle
Physical Form Needles or Powder
Assay Percent Range 99+%
Linear Formula (CHOHCOONa)2·2H2O
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: insolubel in organic solvents
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
Formula Weight 230.08
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material L(+)-Tartaric acid, disodium salt, dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.