missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Tartaric Acid, TRC

Product Code. 30133103
Change view
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133103 250 g 250g
17933735 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133103 Supplier Toronto Research Chemicals Supplier No. TRCT007630250G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4 H6 O6
Molecular Weight (g/mol) 150.09
Synonym Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, Butanedioic acid, 2,3-dihydroxy- [R-(R*,R*)]-, Tartaric acid, L-(+)- (8CI), (2R,3R)-2,3-Dihydroxybutanedioic acid, (+)-(2R,3R)-Tartaric acid, (+)-(R,R)-Tartaric acid, (+)-L-Tartaric acid, (+)-Tartaric acid, (2R,3R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid, (2R,3R)-Tartaric acid, (R,R)-(+)-Tartaric acid, (R,R)-Tartaric acid, 1,2-Dihydroxyethane-1,2-dicarboxylic acid, 2,3-Dihydroxybutanedioic acid, 2,3-Dihydroxysuccinic acid, Dextrotartaric acid, Dihydroxysuccinic acid, E 334, L-(+)-Tartaric acid, L-Tartaric acid, NSC 62778, Natural tartaric acid, Tartaric acid, Threaric acid, d-Tartaric acid, d-α,β-Dihydroxysuccinic acid
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid
SMILES O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1
Formula Weight 150.0164
Percent Purity >95%
Quantity 250 g
Chemical Name or Material L-(+)-Tartaric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.