missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine, 99%

Product Code. 11323197
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323197 25 g 25g
11303197 1 g 1g
11313197 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323197 Supplier Thermo Scientific Alfa Aesar Supplier No. L13707.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20859-02-3 | C6H13NO2 | 131.18 g/mol

L-tert-Leucine is an essential amino acid and makes up one third of our muscle protein. It can be used as a food additive. It is used in the formation of sterols. It is used in the production of cobalt oxazoline palladacycles complex, which acts as a catalyst in the rearrangement of prochiral N-aryl trifluoroacetimidates to allylic amides yielded in significantly higher enantioselectivities with the (S)-(pR)-diasteroisomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-tert-Leucine is an essential amino acid and makes up one third of our muscle protein. It can be used as a food additive. It is used in the formation of sterols. It is used in the production of cobalt oxazoline palladacycles complex, which acts as a catalyst in the rearrangement of prochiral N-aryl trifluoroacetimidates to allylic amides yielded in significantly higher enantioselectivities with the (S)-(pR)-diasteroisomer.

Solubility
Soluble in water and hydrchloric acid.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20859-02-3
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064218
InChI Key NPDBDJFLKKQMCM-SGAVLPGINA-N
Synonym l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
PubChem CID 164608
IUPAC Name (2S)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)[C@H](N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721824
Solubility Information Soluble in water and hydrchloric acid.
Quantity 25 g
Formula Weight 131.18
Optical Rotation +7° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material L-tert-Leucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.