missing translation for 'onlineSavingsMsg'
Learn More

L-Threonic Acid Calcium Salt, TRC

Product Code. 30135167
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135167 100 g 100g
30008538 5 g 5g
30010050 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135167 Supplier Toronto Research Chemicals Supplier No. TRCT404990100G
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70753-61-6
Molecular Formula 2 C4 H7 O5 . Ca
Molecular Weight (g/mol) 310.269
Synonym Butanoic acid, 2,3,4-trihydroxy-, calcium salt (2:1), (2R,3S)-, Butanoic acid, 2,3,4-trihydroxy-, calcium salt (2:1), [R-(R*,S*)]-, Calcium (2R,3S)-2,3,4-trihydroxybutanoate, Calcium L-threonate
IUPAC Name calcium;(2R,3S)-2,3,4-trihydroxybutanoate
SMILES [Ca+2].OC[C@H](O)[C@@H](O)C(=O)[O-].OC[C@H](O)[C@@H](O)C(=O)[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/2C4H8O5.Ca/c2*5-1-2(6)3(7)4(8)9;/h2*2-3,5-7H,1H2,(H,8,9);/q;;+2/p-2/t2*2-,3+;/m00./s1
Formula Weight 310.021
Chemical Name or Material Calcium L-Threonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.