missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, 98%

Product Code. 10001610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10001610 100 g Glass Bottle 100g
10435115 5 g Glass Bottle 5g
10665762 25 g Glass Bottle 25g
10398743 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10001610 Supplier Thermo Scientific Chemicals Supplier No. 138931000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
SMILES CC(O)C(N)C(O)=O

Specifications

Melting Point 255.0°C
Color Yellow
Chloride 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)CO2H
Specific Rotation − 28.30
Sulfate 300ppm max.
Merck Index 15, 9534
Sulfated Ash 0.1 % max.
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: insoluble in most common organic solvents
IUPAC Name 2-amino-3-hydroxybutanoic acid
Specific Rotation Condition −28.30 (20.00°C c=6,H2O)
Quantity 100 g
Formula Weight 119.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.