missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine methyl ester hydrochloride, 98%

Product Code. 15418698
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418698 1 g 1g
15428698 5 g 5g
15438698 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418698 Supplier Thermo Scientific Alfa Aesar Supplier No. H62118.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39994-75-7 | C5H12ClNO3 | 169.61 g/mol

L-Threonine methyl ester hydrochloride is used as a feed and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Threonine methyl ester hydrochloride is used as a feed and food additive.

Solubility
Soluble in water.

Notes
Moisture Sensitive and Hygroscopic. Keep container tightly closed. Store away from water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39994-75-7
Molecular Formula C5H12ClNO3
Molecular Weight (g/mol) 169.61
MDL Number MFCD00037677
InChI Key OZSJLLVVZFTDEY-HJXLNUONSA-N
Synonym l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl
PubChem CID 2734893
IUPAC Name (2S,3R)-3-hydroxy-1-methoxy-1-oxobutan-2-aminium
SMILES Cl.COC(=O)[C@@H](N)[C@@H](C)O

Specifications

Beilstein 3562717
Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 169.61
Sensitivity Hygroscopic; Moisture sensitive
Percent Purity 98%
Chemical Name or Material L-Threonine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.