missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine methyl ester hydrochloride, 98+%

Product Code. 10547113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10547113 5 g Glass Bottle 5g
10482083 1 g Glass Bottle 1g
10500174 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10547113 Supplier Thermo Scientific Acros Supplier No. 376690050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39994-75-7 | C5H12ClNO3 | 169.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39994-75-7
Molecular Formula C5H12ClNO3
Molecular Weight (g/mol) 169.61
MDL Number MFCD00037677
InChI Key OZSJLLVVZFTDEY-HJXLNUONSA-N
Synonym l-threonine methyl ester hydrochloride, methyl l-threoninate hydrochloride, h-thr-ome.hcl, 2s,3r-methyl 2-amino-3-hydroxybutanoate hydrochloride, l-threonine, methyl ester, hydrochloride, l-threonine methyl ester hcl, threonine methyl ester hydrochloride, methyl 2s,3r-2-amino-3-hydroxybutanoate hydrochloride, methyl l-threoninate hcl, h-thr-ome?cl
PubChem CID 2734893
IUPAC Name methyl (2S,3R)-2-amino-3-hydroxybutanoate hydrochloride
SMILES Cl.COC(=O)[C@@H](N)[C@@H](C)O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)C(O)OCH3·HCl
Specific Rotation -9.5° to -10.5° (20°C, 589nm) (c=1, Methanol, 20°C)
Water 1% Max.
Specific Rotation Condition −10° (20°C c=1,methanol)
Quantity 5 g
Formula Weight 169.61
Percent Purity ≥98.0% (on dry substance)
Physical Form Powder
Chemical Name or Material L-Threonine methyleester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.