missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | L-Threonine |
| CAS | 72-19-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C4 H9 N O3 |
| Quantity | 10 mg |
| Synonym | Threonine, (2S,3R)-2-Amino-3-hydroxybutanoic acid, (S)-Threonine, L-Threonine, Threonine','L- (8CI), (S)-Threonine, 2-Amino-3-hydroxybutyric acid, Butanoic acid','2-amino-3-hydroxy-','[R-(R*,S*)]-, L-(-)-Threonine, NSC 16589, NSC 46701, Threonin, [R-(R*,S*)]-2-Amino-3-hydroxybutanoic acid |
| InChI Formula | InChI=1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1 |
| SMILES | C[C@@H](O)[C@H](N)C(=O)O |
| IUPAC Name | (2S,3R)-2-amino-3-hydroxybutanoic acid |
| Molecular Weight (g/mol) | 119.12 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction