missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10502985
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10502985 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10502985 Supplier Fisher BioReagents Supplier No. BP394100
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 255°C
Ammonium 0.02% max.
CAS 72-19-5
pH 5.0 to 6.5
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 99.0 to 101.0 %
Loss on Drying 0.2% max.
Molecular Formula C4H9NO3
Packaging Glass Bottle
Specific Rotation -27.6 to -29.0° (+ or -)
Merck Index 15, 9534
State of Solution (%T) 98.0% min.
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
Alpha Vector Threonine
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
SMILES CC(O)C(N)C(O)=O
IUPAC Name (2S,3R)-2-amino-3-hydroxybutanoic acid
Molecular Weight (g/mol) 119.12
Quantity 100 g
PubChem CID 6288
ChEBI CHEBI:16857
Formula Weight 119.12
Percent Purity 99.0 to 101.0%
Physical Form Powder
Chemical Name or Material L-Threonine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.