missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tryptophan, 99%

Product Code. 11473923
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473923 500 g 500g
11453923 25 g 25g
11463923 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473923 Supplier Thermo Scientific Chemicals Supplier No. A10230.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol

L-tryptophan is an essential amino acid which is necessary for normal growth in infants and for nitrogen balance in adults. It acts as a natural dietary supplement and used as an antidepressant, anxiolytic and sleep aid. It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-tryptophan is an essential amino acid which is necessary for normal growth in infants and for nitrogen balance in adults. It acts as a natural dietary supplement and used as an antidepressant, anxiolytic and sleep aid. It is used as a precursor to niacin, indole alkaloids and serotonin. It acts as an important intrinsic fluorescent probe, which finds to estimate the nature of microenvironment of the tryptophan.

Solubility
Soluble in water, ammonia, hot alcohol, alkali hydroxides and dilute acids. Slightly soluble in acetic acid and ethanol. Insoluble in chloroform and ethyl ether.

Notes
Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 280°C 285°C
Assay Percent Range 99%
Beilstein 86197
Merck Index 14,9797
Solubility Information Soluble in water,ammonia,hot alcohol,alkali hydroxides and dilute acids. Slightly soluble in acetic acid and ethanol. Insoluble in chloroform and ethyl ether.
Quantity 500 g
Formula Weight 204.23
Optical Rotation −31° (c=1 in water)
Percent Purity 99%
Chemical Name or Material L-Tryptophan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.