missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, ≥99%, MP Biomedicals™

Product Code. 11453581 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453581 1 kg 1kg
11423581 25 g 25g
11433581 100 g 100g
11443581 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11453581 Supplier MP Biomedicals Supplier No. 0210315191
  • ONLINE EXCLUSIVE PRICE
    1325.00 €
    1255.80 € / 1kg
    Save 69.20 €
    5% Off
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-Tryptophan is an amino acid precursor of serotonin and melatonin

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 1 kg
Formula Weight 204.22
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material L-Tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.