missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Benzyl Ester 4-Toluenesulfonate, TRC

Product Code. 30454146
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454146 Supplier Toronto Research Chemicals Supplier No. TRCT966383500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Tyrosine Benzyl Ester 4-Toluenesulfonate
CAS 53587-11-4
Molecular Formula C16H17NO3 . C7H8O3S
Quantity 500 mg
Synonym L-Tyrosine','phenylmethyl ester','4-methylbenzenesulfonate (1:1), L-Tyrosine','phenylmethyl ester','4-methylbenzenesulfonate (salt) (9CI), (S)-Tyrosine benzyl ester tosylate, Benzyl L-tyrosinate tosylate, L-Tyrosine benzyl ester 4-methylbenzenesulfonate, L-Tyrosine benzyl ester p-toluenesulfonate, Tyrosine benzyl ester tosylate, Tyrosine benzyl ester tosylate (salt)
InChI Formula InChI=1S/C16H17NO3.C7H8O3S/c17-15(10-12-6-8-14(18)9-7-12)16(19)20-11-13-4-2-1-3-5-13;1-6-2-4-7(5-3-6)11(8,9)10/h1-9,15,18H,10-11,17H2;2-5H,1H3,(H,8,9,10)/t15-;/m0./s1
SMILES Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](Cc2ccc(O)cc2)C(=O)OCc3ccccc3
IUPAC Name benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid
Molecular Weight (g/mol) 443.51
Formula Weight 443.1403
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.