missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine disodium salt dihydrate

Product Code. 15484289
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484289 500 g 500g
15474289 100 g 100g
15494289 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15484289 Supplier Thermo Scientific Chemicals Supplier No. J61770.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 122666-87-9 | C9H13NNa2O5 | 261.19 g/mol

Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122666-87-9
Molecular Formula C9H13NNa2O5
Molecular Weight (g/mol) 261.19
MDL Number MFCD03840558
InChI Key WFTCFVUQORELJZ-CMIWSEPMNA-L
Synonym tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate
PubChem CID 71587456
SMILES O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Specifications

Color Brown to White
Solubility Information Soluble in water.
IUPAC Name disodium;(2S)-2-amino-3-(4-oxidophenyl)propanoate;dihydrate
Quantity 500 g
Formula Weight 261.18 (225.15 Anhydrous)
Optical Rotation −13° (c=4 in Water)
Sensitivity Very Hygroscopic
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Tyrosine disodium salt dihydrate
compliance-icons
Product Identifier
  • L-Tyrosine disodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : L-Tyrosine, sodium salt, hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.