missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11458840
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458840 100 g 100g
11448840 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458840 Supplier Thermo Scientific Alfa Aesar Supplier No. A18358.22
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a medical and organic intermediate and as an important amino protective agent. It is also used to introduce t-Boc protect gene.

Solubility
Soluble in water (3.685e+005 mg/L @ 25°C (est.)).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4089-07-0
Molecular Formula C11H16ClNO3
Molecular Weight (g/mol) 245.70
MDL Number MFCD00063047
InChI Key BQULAXAVRFIAHN-OXZMBGLCNA-N
Synonym l-tyrosine ethyl ester hydrochloride, h-tyr-oet.hcl, ethyl l-tyrosinate hydrochloride, unii-fap58daz1e, s-ethyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, h-tyr-oet hcl, tyrosine ethyl ester hydrochloride, fap58daz1e, tyrosine, ethyl ester, hydrochloride, l-tyrosine, ethyl ester hydrochloride
PubChem CID 2724939
IUPAC Name ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
SMILES Cl.CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 166°C to 168°C
Beilstein 4725904
Solubility Information Soluble in water (3.685e+005mg/L @ 25°C (est.)).
Quantity 100 g
Formula Weight 245.71
Optical Rotation −6.8° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Tyrosine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.