missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine methyl ester, 98%

Product Code. 15448658
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448658 100 g 100g
15438658 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448658 Supplier Thermo Scientific Alfa Aesar Supplier No. H62069.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1080-06-4 | C10H13NO3 | 195.22 g/mol

L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tyrosine methyl ester is used as a dehydroepiandrosterone as a sulfate acceptor

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1080-06-4
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002392
InChI Key MWZPENIJLUWBSY-VIFPVBQESA-N
Synonym l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 70652
ChEBI CHEBI:17215
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Spezifikation

Melting Point 134°C to 136°C
Beilstein 2372626
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material L-Tyrosine methyl ester

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.