missing translation for 'onlineSavingsMsg'
Learn More

L-Valine Methyl Ester Hydrochloride, TRC

Product Code. 30448942
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30448942 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30448942 Supplier Toronto Research Chemicals Supplier No. TRCV09432510G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Valine Methyl Ester Hydrochloride
CAS 6306-52-1
Molecular Formula C6 H13 N O2 . Cl H
Quantity 10 g
Synonym L-Valine','methyl ester','hydrochloride (1:1), L-Valine','methyl ester','hydrochloride (9CI), Valine','methyl ester','hydrochloride','L- (6CI,7CI,8CI), (S)-Valine methyl ester hydrochloride, Methyl (S)-2-amino-3-methylbutanoate hydrochloride, Methyl (S)-valinate hydrochloride, Methyl L-valinate hydrochloride, Methyl valinate hydrochloride, NSC 197198, NSC 22920, Valine methyl ester hydrochloride
InChI Formula InChI=1S/C6H13NO2.ClH/c1-4(2)5(7)6(8)9-3;/h4-5H,7H2,1-3H3;1H/t5-;/m0./s1
SMILES Cl.COC(=O)[C@@H](N)C(C)C
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 167.63
Formula Weight 167.0713
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.