missing translation for 'onlineSavingsMsg'
Learn More

L-Valine (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10477773
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10477773 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10477773 Supplier Fisher BioReagents Supplier No. BP397100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 315°C
Ammonium 0.02% max.
CAS 72-18-4
Arsenic (As) 1.0ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Identification Pass Test
Assay Percent Range 98.5 to 100.5 %
Loss on Drying 0.2% max.
Other Amino Acids Not Detected
Molecular Formula C5H11NO2
Packaging Glass Bottle
MDL Number MFCD00064220
Specific Rotation 27.6 to 29.0° (+ or -)
Merck Index 15, 10095
State of Solution (%T) 98.0% min.
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
Alpha Vector Valine
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Density 1.230g/cm³
SMILES CC(C)[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
Molecular Weight (g/mol) 117.15
Quantity 100 g
PubChem CID 6287
ChEBI CHEBI:16414
Formula Weight 117.08
Percent Purity 98.5 to 100.5%
Physical Form Powder
Chemical Name or Material L-Valine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.