missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Xylose, 99%

Product Code. 11477244
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11477244

Brand: Thermo Scientific Chemicals B21622.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 609-06-3 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-06-3
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00151096
InChI Key PYMYPHUHKUWMLA-UHFFFAOYNA-N
Synonym l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
PubChem CID 95259
ChEBI CHEBI:17979
SMILES OCC(O)C(O)C(O)C=O

Specifications

Melting Point 147°C to 151°C
Density 1.525
Quantity 5 g
Beilstein 1723080
Sensitivity Hygroscopic
Optical Rotation −20° (c=10 in Water, 10h)
IUPAC Name (2S,3R,4S)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material L-(-)-Xylose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.