missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Packs with Screw-On Lid

Product Code. 17752308
Change view
Click to view available options
Capacity (Metric):
75.7 L
113 L
217 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17752308 75.7 L Each
17742308 113 L Each
17762308 217 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17752308 Supplier Eagle™ Supplier No. EDR16500YL
  • ONLINE EXCLUSIVE PRICE
    115.26 €
    97.14 € / Pack of 1
    Save 18.12 €
    15% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eagle Lab Packs are ideal for safe storage, handling and transportation of hazardous materials, spill containment and clean-up of hazardous materials, original shipping container, bio waste disposal and incineration.

  • Eagle Lab Pack Drums are constructed of Yellow blow-molded, high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, weather and chemical resistant, and screw-on lid
  • Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
  • Lab Packs/Open Head Drums- for use with smaller non-damaged bottles, cans, carboys and 5-gallon pails of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS

Specifications

Capacity (Metric) 75.7 L
Color Yellow
For Use With (Application) Safe Storage, Handling and Transportation of Hazardous Materials
Material Polyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.