missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Avidin biotin Blocking Solution

Product Code. 12683937 Shop All Epredia Products
Change view
Click to view available options
Capacity:
15 mL (Biotin)
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
12683937 15 mL (Biotin) 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12683937 Supplier Epredia™ Supplier No. TA015BB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Block nonspecific staining due to endogenous biotin in immunohistochemical staining with Epredia™ Lab Vision™ Avidin Biotin Blocking Solution

  • Blocks nonspecific staining due to endogenous biotin during the IHC detection of antigens

Storage Conditions: Store between 4° to 6°C. Components stable for up to 18 months.
TRUSTED_SUSTAINABILITY

Specifications

Capacity 15 mL (Biotin)
Content And Storage Store at 4° to 6°C

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.