missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ biotinylated Goat anti-Polyvalent (Ready-To-Use)

Product Code. 12674047 Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
125mL
Unit Size:
125mL
60mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12674047 60 mL 60mL
12624057 125mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12674047 Supplier Epredia™ Supplier No. TP060BN

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Employ a labeled streptavidin-biotin immunoenzymatic antigen detection system with the Epredia™ Lab Vision™ Biotinylated Goat anti-Polyvalent (Ready-To-Use).

This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate-chromogen.
  • Species: Goat

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Conjugate: Biotin

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin
Content And Storage 2° to 8°C
Product Type Anti-Polyvalent
Quantity 60 mL
Species anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.