Learn More
Epredia™ Lab Vision™ Caspase 6 (Mch 2) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and Azide
Ensure accurate, reproducible results in western blotting and immunoprecipitation procedures with Epredia™ Caspase 6 (Mch 2) Ab-3, Mouse Monoclonal Antibody.
248.00 € - 945.00 €
Specifications
| Antigen | Caspase 6 (Mch 2) Ab-3 |
|---|---|
| Clone | 6CSP03; same as 14.1.190 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|
12639056
|
Epredia™
MS-1126-P0 |
100 μL | 200μg/mL | ||||||
|
12649056
|
Epredia™
MS-1126-P1 |
500 μL | 200μg/mL | ||||||
|
12659056
|
Epredia™
MS-1126-P1ABX |
100 μL | 1mg/mL | ||||||
|
12669056
|
Epredia™
MS-1126-PABX |
200 μL | 1mg/mL | ||||||
Description
Caspases play a major role in the transduction of the apoptotic signal and execution of apoptosis in mammalian cells. Caspase 3 (CPP32) and Caspase 6 (Mch2) are the major active caspases in apoptotic cells, and are activated in response to distinct apoptosis-inducing stimuli in all cell lines analyzed. Both CPP32 and Mch2 are present in apoptotic cells as multiple active species. Caspase-6 cleaves nuclear mitotic apparatus protein (NuMA) and mediates the shrinkage and fragmentation of nuclei.Host Species: Mouse
Clone: 6CSP03; same as 14.1.190
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Caspase 6 prodomain
Immunogen: Recombinant human caspase 6 prodomain
Molecular Weight: 34 kDa
Positive Control: Jurkat cells
Cellular Localization: Cytoplasmic
Recommended for:
- Western Blotting
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
| Caspase 6 (Mch 2) Ab-3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| Recombinant human caspase 6 prodomain | |
| Primary |
| 6CSP03; same as 14.1.190 | |
| Monoclonal | |
| Concentrated | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.