missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lab Vision™ DAB Plus Substrate Staining System

Product Code. 12683957 Shop All Epredia Products
Click to view available options
Quantity:
125 mL Substrate, 4mL Chromogen
Unit Size:
125mL
This item is not returnable. View return policy
DAB Plus Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Plus Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.

Product Code. 12683957

Brand: Thermo Scientific™ TA125HDX

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
DAB Plus Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Plus Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.

Take advantage of 3,3' Diaminobenzidine (DAB), a widely used chromogen for immunohistochemical staining and immunoblotting.

Take advantage of 3,3' Diaminobenzidine (DAB), a widely used chromogen for immunohistochemical staining and immunoblotting with the Thermo Scientific™ Lab Vision™ DAB Plus Substrate Staining System is ideal for automated stainers.

In the presence of peroxidase enzyme, DAB produces a brown precipitate that is insoluble in alcohol.
  • Supplied in a two-component form consisting of a DAB Plus Chromogen and DAB Plus Substrate
  • Standard working dilution is 20μL of DAB Plus Chromogen per 1mL of DAB Plus Substrate, although the ratio can be adjusted as desired
  • Use of liquid components reduces the risk of dust inhalation
  • Prepare only what is needed, eliminating the waste associated with reagents in tablet form
TRUSTED_SUSTAINABILITY

Specifications

Chromagen Quantity 4mL
Chromagen Type DAB Plus
Quantity 125 mL Substrate, 4mL Chromogen
Substrate Type DAB Substrate
Storage Requirements Store at 2° to 8°C
Substrate Quantity 125mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.