Learn More
Epredia™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Rabbit Polyclonal Antibody with BSA and azide
Epredia™ MMP-9 (92kDa Collagenase IV), Rabbit Polyclonal Antibody provides accurate, reproducible results.
131.58 €
Specifications
Antigen | MMP-9 (92kDa Collagenase IV) |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12663497
|
Epredia™
RB-9234-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Metallomatrix proteases (MMPs) are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. This antibody reacts with both latent and active MMP-9.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Guinea Pig. Does not react with mouse, rat or bovine. Others not tested.
Epitope: Near C-terminus
Immunogen: A synthetic peptide derived from near C-terminal of human MMP-9 protein
Molecular Weight: 92kDa (proform) and ∽86kDa (active form)
Positive Control: Placenta, or Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
MMP-9 (92kDa Collagenase IV) | |
Polyclonal | |
Rabbit | |
Proteases And Inhibitors | |
A synthetic peptide derived from near C-terminal of human MMP-9 protein | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Guinea Pig | |
IgG |