missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ MultiVision Polymer Detection System: anti-Mouse-HRP and anti-Rabbit-AP

Product Code. 12693987 Shop All Epredia Products
Click to view available options
Quantity:
12 mL
Unit Size:
12mL
This item is not returnable. View return policy

Product Code. 12693987

Brand: Epredia™ TL012MHRA

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A simple and robust one-step polymer detection system for visualization of two antigens simultaneously in blue and red.

MultiVision AP and HRP polymers are innovative, patented technologies consisting of smaller amino acid polymer subunits that minimize conflicts in binding the target protein. The smaller polymer allows for decreased binding conflicts, resulting in more consistent staining and better signal amplification.
  • A primary antibody cocktail of choice may be used in conjunction with ready-to-use MultiVision polymer cocktail, composed of anti-mouse/ horseradish peroxidase (HRP) plus anti-rabbit/alkaline phosphatase (AP)
  • Cocktail is specially formulated from MultiVision polymers that provide increased sensitivity, time-savings, and detection simplicity
  • Yields higher sensitivity and antibody efficiency as well as better signal-to-noise ratios
  • Biotin-free, eliminating background staining
  • Antigen Specificity: anti-Mouse IgG (H+L) and anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase and Phosphatase

  • Chromogen/Substrate: LVBlue/ LVRed

Includes:

TL-012-MHRA: 1 MultiVision anti-rabbit/AP + anti-mouse/HRP polymer (cocktail) (TL-012-RAM), 1 Ultra V block (TA-012-UB), 1 Hydrogen Peroxidase Block (TA-012-HP), 1 LVBlue AP Buffer (TA-075-TMM), 1 LVBlue Chromogen Vial 1 (TA-001-AB1), 1 LVBlue Chromogen Vial 2 (TA-001-AB2), 1 LVBlue Chromogen Vial 3 (TA-001-AB3), 1 LVRed Chromogen Vial 1 (TA-004-HR1), 1 LVRed Chromogen Vial 2 (TA-002-HR2), 1 LVRed Chromogen Vial 3 (TA-002-HR3), 1 LVRed Chromogen Vial 4 (TA-002-HR4), 1 Mounting Media (TA-005-UA)

Note:

MultiVision Polymer Detection System may be employed with any primary antibody cocktail to visualize the presence of two proteins simultaneously. After the specific antibodies are bound to their respective antigens in tissue sections, the rabbit and mouse primary antibodies are located by a specially formulated cocktail of secondary antibody polymers: anti-mouse/ horseradish peroxidase (HRP) + anti-rabbit/alkaline phosphatase (AP). In general, the amino acid polymer is conjugated to AP or HRP enzymes and the Fab fragments of goat anti-rabbit or goat anti-mouse immunoglobulins. The polymer complex is then visualized with first the LVBlue chromogen for AP activity and after washing with the LVRed chromogen for HRP activity. A weak hematoxilin nuclear counterstain is not recommended.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 12 mL
Includes TL-012-MHRA: 1 MultiVision anti-rabbit/AP + anti-mouse/HRP polymer (cocktail) (TL-012-RAM), 1 Ultra V block (TA-012-UB), 1 Hydrogen Peroxidase Block (TA-012-HP), 1 LVBlue AP Buffer (TA-075-TMM), 1 LVBlue Chromogen Vial 1 (TA-001-AB1), 1 LVBlue Chromogen Vial 2 (TA-001-AB2), 1 LVBlue Chromogen Vial 3 (TA-001-AB3), 1 LVRed Chromogen Vial 1 (TA-004-HR1), 1 LVRed Chromogen Vial 2 (TA-002-HR2), 1 LVRed Chromogen Vial 3 (TA-002-HR3), 1 LVRed Chromogen Vial 4 (TA-002-HR4), 1 Mounting Media (TA-005-UA)
Chromagen LVBlue/
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Storage Requirements Store at 2° to 8°C. Stable for up to 12 months. Store chromogens away from light.
Enzyme Peroxidase, Phosphatase
Product Line Lab Vision
Substrate LVRed
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.