missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ PT Module™ Deparaffinization and Heat-Induced Epitope Retrieval Solutions (100X), Citrate buffer, pH 6

Product Code. 12633967 Shop All Epredia Products
Change view
Click to view available options
Quantity:
125mL
250mL
pH:
6
8
9
Unit Size:
125mL
250mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH unitSize
12633967 125mL 6 125mL
12643967 125mL 8 125mL
12663967 125mL 9 125mL
12633987 250mL 6 250mL
12643987 250mL 8 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12633967 Supplier Epredia™ Supplier No. TA125PM1X

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform Heat Induced Antigen Retrieval (HIER) on formalin fixed paraffin-embedded tissue sections with Epredia™ Lab Vision™ PT Module™ Epitope Retrieval Solutions.

  • 100X stock solution
  • Dilute 100-fold with distilled water before use
  • Designed for use with Pretreatment Module™
  • Available in Citrate, EDTA or Tris-HCl at various pH levels

Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives; for periods longer than three months, store at 2°-8°C.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Dilution 100X
Product Type HIER
Quantity 125mL
For Use With (Application) Immunohistochemistry
pH 6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.