missing translation for 'onlineSavingsMsg'
Learn More

Lab Vision™ Tris Buffered Saline and Tween™ 20 (20X), Epredia™

Product Code. 12880375 Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
1000 mL
Unit Size:
1 L
125mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12880375 1000 mL 1 L
12633977 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12880375 Supplier Epredia Supplier No. TA999TT
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Take advantage of a wash buffer for immunolabeling techniques with the Utilize Epredia™ Lab Vision™ Tris-Buffered Saline and Tween™ 20 (20X).

  • For in vitro diagnostic use
  • Tween 20 is added to promote more effective washings, resulting in decreased nonspecific background staining
  • Tween 20 aids reagent spreading on slides

Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBS with Tween (TBST)
Concentration 20X
For Use With (Application) Anatomical Pathology
Quantity 1000 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.