Learn More
Labconco™ Atmospure Tubing Kit
Tubing Kits connect AtmosPure Re-Gen Gas Purifier to glove box
Brand: Labconco™ 5441801
Additional Details : Weight : 9.10000kg
Description
AtmosPure Circulation Tubing Kit includes the plumbing to make the connection from the Labconco AtmosPure Re-Gen Gas Purifier to the glove box. Includes 3/4 in. ID neoprene-lined flexible hose. For use with all double-width glove boxes.Specifications
Double Width Glove Box | |
20 lb. | |
9.1 Kg | |
Atmospure Flex Tubing Kit, Double |