missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Clear Stoppering Chamber

Product Code. 10183432 Shop All Labconco Corporation Products
Change view
Click to view available options
Electrical Requirements:
230 V, 50/60 Hz, International
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Unit Size
10183432 230 V, 50/60 Hz, International Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10183432 Supplier Labconco™ Supplier No. 7868030
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clear Stoppering Chamber for use with 6 L, 12 L and 18 L Freezone™ Dryers.

  • Allows flasks attachment with freeze dry system or contain samples in bulk

Specifications

Type Clear Stoppering Chamber
No. of Shelves 1
Temperature Range (Metric) Up to 40°C
Material Acrylic
For Use With (Application) To provide economical means of stoppering serum bottles
For Use With (Equipment) 6 L, 12 L and 18 L FreeZone™ Dryers
Diameter (English) 13.5 in.
Diameter (Metric) 34 cm
Height (English) 23 in.
Height (Metric) 58.4 cm
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz, International
Frequency 50/60 Hz
Weight (English) 30 lb.
Weight (Metric) 13.6 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.