missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Mini Stoppering Chamber

Product Code. 15342272 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
FreeZone™ Freeze Dry System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15342272 FreeZone™ Freeze Dry System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15342272 Supplier Labconco™ Supplier No. 7569000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

FreeZone Freeze Dry System to attach Mini Stoppering Chamber via port

For lyophilizing samples in small serum vials with split stoppers

  • For use with a FreeZone™ Freeze Dry System
  • Accommodates 48 serum vials of 2 mL and 8 serum vials of 30 mL
  • After lyophilization, vials can be stoppered while still under vacuum
  • Stoppering travel distance: 0.5 in. (1.3 cm)
  • For vials ranging in height from 1.38 to 2.48 in. (3.5 to 6.3 cm)
  • Comes with tray for loading and unloading of samples

Specifications

Type Mini Stoppering Chamber
No. of Shelves 0
Material Acrylic
For Use With (Application) For lyophilizing samples in small serum vials with split stoppers
For Use With (Equipment) FreeZone™ Freeze Dry System
Includes 17 in. of clear pvc tubing
Width (English) 12 in.
Width (Metric) 30.4 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Weight (English) 15 lb.
Weight (Metric) 6.8 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.