missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AHDC1 Full-length ORF (AAH14394.2, 1 a.a. - 399 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16108702
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16108702 10 μg 10µg
16118702 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16108702 Supplier Abnova™ Supplier No. H00027245P01.10ug
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sequence: MMDWNEASSAPGYNWNQSVLFQSSSKPGRGRRKKVDLFEASHLGFPTSASAAASGYPSKRSTGPRQPRGGRGGGACSAKKERGGAAAKAKFIPKPQPVNPLFQDSPDLGLDYYSGDSSMSPLPSQSRAFGVGERDPCDFIGPYSMNPSTPSDGTFGQGFHCDSPSLGAPELDGKHFPPLAHPPTVFDAGLQKAYSPTCSPTLGFKEELRPPPTKLAACEPLKHGLQGASLGHAAAAQAHLSCRDLPLGQPHYDSPSCKGTAYWYPPGSAARSPPYEGKVGTGLLADFLGRTEAACLSAPHLASPPATPKADKEPLEMARPPGPPRGPAAAAAGYGCPLLSDLTLSPVPRDSLLPLQDTAYRYPGFMPQAHPGLGGGPKSGFLGPMAEPHPEDTFTVTSL

Specifications

Accession Number AAH14394.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27245
Molecular Weight (g/mol) 68.1kDa
Name AHDC1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CL23945/DJ159A19.3/RP1-159A19.1
Common Name AHDC1
Gene Symbol AHDC1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.