Learn More
Labconco™ Protector Glove Box Accessory Kits
Product Code. 16649183 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Pressure Relief Kit
Unit Size:
Each
Product Code. Product Type Unit Size
16649183 Pressure Relief Kit Each
1 options
Product Code. 16649183 Supplier Labconco™ Supplier No. 5441403
  • ONLINE EXCLUSIVE PRICE
    871.00 €
    858.96 € / Pack of 1
    Save 12.04 €
    1% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Kits provided added functionality options for Protector glove boxes
Kit Components
  • 5 in. diameter round HEPA filter (99.9% efficient on 0.3 micron particulates)
  • Plastic hose barbs (2)
  • 10 ft. of 3/4 in. ID neoprent tubing
Specifications
For Use With (Equipment) Protector Glove Boxes
Includes HEPA filter, hose barb connectors and 10 ft. of 0.75 in. ID neoprene tubing for attachment to the pressure relief bubbler exhaust
Product Type Pressure Relief Kit