missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Standard Storage Cabinet

Product Code. 15189705 Shop All Labconco Corporation Products
Change view
Click to view available options
Width (English):
36 in.
No. of Doors:
2
Door Type:
Manual Close
Weight (English):
180 lb.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) No. of Doors Door Type Weight (English) Unit Size
15189705 36 in. 2 Manual Close 180 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15189705 Supplier Labconco™ Supplier No. 9900100
  • ONLINE EXCLUSIVE PRICE
    1460.00 €
    1284.48 € / Pack of 1
    Save 175.52 €
    12% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

  • Support loads up to 800 lb. (363kg)
  • Durable epoxy-coated steel construction
  • Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
  • Manual-closing, non-locking door(s)

Order Info

Work surface and interior shelf kits are sold separately

Specifications

Product Type Storage Cabinet
Color White
Door Type Manual Close
For Use With (Application) Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Height (English) 35.5 to 36.75 in.
Height (Metric) 90.17 to 93.34 cm
Includes Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
Length (English) 22 in.
Load Bearing Capacity (English) 800 lb.
Load Bearing Capacity (Metric) 362.87 kg
No. of Doors 2
Surface Coating Epoxy
Weight (English) 180 lb.
Weight (Metric) 81.64 kg
Width (English) 36 in.
Width (Metric) 91.44 cm
Material Epoxy-coated Steel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.