missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Label Printer Hard Case

Product Code. 17862609 Shop All Brady Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17862609 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17862609 Supplier Brady™ Supplier No. 176448
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect, transport and store your M511 Label Printer, plus printer accessories and label cartridges.

Protect, transport, and store your M511 Label Printer, plus printer accessories and label cartridges.

  • Protects your printer from impacts and dust.
  • Keeps printer, accessories and label cartridges organized and on hand.
  • Durable hard case allows for easy mobility.

Specifications

Product Type Hard Case
Color Blue
For Use With (Equipment) Brady Label Printers
Material Plastic
Length (English) 6.5 in.
Width (English) 17 in.
Height (English) 17.5 in.
Description M511 Printer Hard Case
Includes (1) Hard Case
For Use With (Application) Asset Tracking;Circuit Board Labeling;Component and Equipment Labeling;Data and Telecommunications Labeling;Electrical Labeling;Facility Identification;General Identification;Inventory and Inspection Labeling;Laboratory Labeling;Lean and 5S Labeling;Safety Identification;Warehouse Marking;Wire and Cable Labeling
Alternate Part Number M511-HC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.