Learn More
Hirschmann™ Class AS AR-Glas Volumetric Pipettes
Product Code. 10625562
Change view
Click to view available options
Volume (Metric):
4 mL
Unit Size:
Pack of 6
Product Code. Volume (Metric) Unit Size
10625562 4 mL Pack of 6
1 options
Product Code. 10625562 Supplier Hirschmann™ Supplier No. 1341150
  • ONLINE EXCLUSIVE PRICE
    48.00 €
    39.16 € / Pack of 6
    Save 8.84 €
    18% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann™ AR™ Glass Volumetric Pipettes, Class AS come with blue graduation.
  • Acc. to DIN EN ISO 648
  • One mark, blue graduation
  • Conformity certified, with dated batch identification
Specifications
Class AS
Graduated Yes
Graduation Color Blue
Quantity 6/Pk.
Volume (Metric) 4 mL
Type Labeled Pipet